Gas and vapour cartridge filters for masks, 6000 series
供应商: 3M
MMMA6001EA 0 CNY
MMMA6001 MMMA6006 MMMA6002 MMMA6003 MMMA6004
Gas and vapour cartridge filters for masks, 6000 series
呼吸器 可重复使用的呼吸器 Respirator Filters
The 3M™ cartridge filters 6000 Series attach to 3M™ half and full face respirators to provide gas and vapour protection.
订购须知 Two cartridges per package/order.