4,4'-二甲氧基二苯甲酮 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Bis(4-methoxyphenyl)methanone
161740250
161741000
ACRO161740250EA
0
CNY
ACRO161740250
ACRO161741000
4,4'-二甲氧基二苯甲酮 97%
4,4'-二甲氧基二苯甲酮
分子式:
C₁₅H₁₄O₃ 分子量: 242.27 g/mol 溶点: 141…146 °C 存储条件: Ambient |
MDL编号:
MFCD00008404 CAS号: 90-96-0 EINECS: 202-028-0 |
产品技术规格
Appearance | Yellow to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 141 to 146 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...