VWR®, Calibration Weights, Class F1
供应商: VWR Collection
VWRI611-8028EA 4873 CNY
VWRI611-8028 VWRI611-8029 VWRI611-8033 VWRI611-8035 VWRI611-8036 VWRI611-8037 VWRI611-8038 VWRI611-8201 VWRI611-8202 VWRI611-8203 VWRI611-8204 VWRI611-8205 VWRI611-8206 VWRI611-8207 VWRI611-8208 VWRI611-8209 VWRI611-8210 VWRI611-8211 VWRI611-8212 VWRI611-8213 VWRI611-8214 VWRI611-8215 VWRI611-8216 VWRI611-8217 VWRI611-8218 VWRI611-8219 VWRI611-8220 VWRI611-8221 VWRI611-8222 VWRI611-8039 VWRI611-8040 VWRI611-8041 VWRI611-8042 VWRI611-8049 VWRI611-8058 VWRI611-8071 VWRI611-8093 VWRI611-8231 VWRI611-8232 VWRI611-8233 VWRI611-8234 VWRI611-8235 VWRI611-8236 VWRI611-8237 VWRI611-8238 VWRI611-8239 VWRI611-8240 VWRI611-8241 VWRI611-8242 VWRI611-8243 VWRI611-8244 VWRI611-8245 VWRI611-8246 VWRI611-8247 VWRI611-8248 VWRI611-8249 VWRI611-8250 VWRI611-8251 VWRI611-8252 VWRI611-8094
VWR®, Calibration Weights, Class F1
称量产品 校准砝码
Weights made from anti-magnetic, polished stainless steel. Suitable for medium resolution precision electronic balances. Weights have adjustment cavities for weights of more than 20 g.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Conform to OILM R111 recommendations.
Certifications: Conform to OIML R111 recommendations.
订购须知 All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. A range of weight sets are also available - please search for product 'Calibration weight sets, Class F1'.
Packaging: Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.