Core samplers, QuickPicker
供应商: BURKLE
5322-1300 5322-0300
BURK5322-1300EA 0 CNY
BURK5322-1300 BURK5322-0300
Core samplers, QuickPicker
采样器 固体取样器
Made of stainless steel or PP, transparent, sample bottles made of PE. The QuickPicker is appropriate when bulk goods are withdrawn directly from sacks, bags or open containers. The sample is filled directly into sample bottles. Sampling depth 300 mm, Ø 25 mm, length 500 mm, 75 ml volume.
  • Removable tip
  • Easy to handle and clean
  • No edges or crevices, no residual sample material
Delivery information: QuickPicker is delivered complete with two sample bottles and a cleaning brush.