Zone samplers
供应商: BURKLE
5317-1150 5317-1058 5317-1085 5316-0055 5316-0085 5319-1085 5319-1150 5316-1055 5316-1085 5317-2055 5317-2150 5317-2085 5319-1250 5316-1150 5316-3150 5316-0150 5316-3055 5316-2055 5316-3085 5316-2085 5316-2150 5319-2085 5319-2150 5319-2250
BURK5317-1150EA 0 CNY
BURK5317-1150 BURK5317-1058 BURK5317-1085 BURK5316-0055 BURK5316-0085 BURK5319-1085 BURK5319-1150 BURK5316-1055 BURK5316-1085 BURK5317-2055 BURK5317-2150 BURK5317-2085 BURK5319-1250 BURK5316-1150 BURK5316-3150 BURK5316-0150 BURK5316-3055 BURK5316-2055 BURK5316-3085 BURK5316-2085 BURK5316-2150 BURK5319-2085 BURK5319-2150 BURK5319-2250
Zone samplers
采样器 固体取样器
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain. Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizes and quantities.
  • Easy to clean, removable tip
  • Colour coding (Multi and Uno samplers)
  • Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
  • 3 lengths from 550 mm to 1500 mm, Jumbo up to 2500 mm
  • Samplers have smallest possible gap between inner and outer tubes