Cartridge burner
供应商: USBECK LABORGERATE
1420 1425
USBE1420EA 0 CNY
USBE1420 USBE1425
Cartridge burner
加热器 煤气灯
Stainless steel burner for propane/butane mix. For temperatures up to 1400 °C.
  • Air regulation and needle valve
Ø×H: 105×215 mm
Ø×H: 105×215 mm
Delivery information: Supplied with cartridge, refill as accessory.