Large horizontal gel electrophoresis systems, Owl™ A1
供应商: THERMO OWL SCIENTIFIC
A1 A1-BP
OWLAA1EA 0 CNY
OWLAA1 OWLAA1-BP
Large horizontal gel electrophoresis systems, Owl™ A1
电泳系统 水平凝胶电泳系统
The Owl™ A1 large gel system is a large format device with a small footprint, ideal for detailed DNA/RNA analysis. The unit runs from 8 to 96 samples on one gel and is available with or without buffer exchange ports.
  • Each side of the UV-transmissible gel tray is fitted with gasketed end gates, which provides a leak-proof seal without tape
  • Built-in levelling ensures the casting of flat, uniformly thick gels
  • Multi-channel pipette compatible combs speed up sample transfer and reduce the chance of sample order error
  • Comb slots on the UVT gel tray are positioned so that up to 10 sample sets of equal lengths can be arranged on the same gel
Delivery information: Each system includes buffer chamber with three point levelling base, SuperSafe lid with attached power leads, UV-transmissible gel tray with end gates, and three combs: 1,5 mm thick, 12-, 16- and 20-well.