Baths and lids, plastic, P series
供应商: GRANT INSTRUMENTS
P12 P18 P5
GRANP12EA 0 CNY
GRANP12 GRANP18 GRANP5
Baths and lids, plastic, P series
浴锅 非搅拌水浴锅
A range of cost-effective plastic baths, for use with all Optima thermostats.
  • Temperature range ambient +5 up to 99 °C
  • Rugged construction, double-walled for rigidity
  • Easy to clean
订购须知 All baths include bridge plate for controller mounting. Lids are recommended when working above 60 °C. Please order lids separately.
Caution: * RT = Ambient