Semi-micro and analytical balances, Excellence XA and XS series
供应商: METTLER TOLEDO
11106009 11106000 11106015 11106003 11106006 11106012
METB11106009EA 0 CNY
METB11106009 METB11106000 METB11106015 METB11106003 METB11106006 METB11106012
Semi-micro and analytical balances, Excellence XA and XS series
天平
The XS and XA analytical balances provide fast and accurate weighing with flexibility. This is achieved with the introduction of SmartGrid technology, with the gridded weighing pan supported at the rear of the weighing chamber. Therefore the draught shield and drip tray can be completely, quickly and easily removed. The draught shield can be set to open the right hand door with the left slider and vice versa making loading more efficient. The design also features a large backlit touch screen that facilitates direct access to all functions and a standard range of applications, that include simple weighing, percentage weighing, piece counting and statistics. Software can be downloaded from the internet via RS232 connection.
  • SmartGrid and ErgoClips for fast results and secure sample handling
  • SmartScreen touch screen with SmartTrac graphical weighing-in guide on XA models
  • FACT fully automatic time and temperature controlled internal adjustment and linearisation
  • Adjustable and detachable display gives flexibility in situating the units
In addition to the benefits features listed previously the wider range of XS balances also feature:
More advanced applications including density determinations, formula weigh and totalise and calculate coefficients. GLP compliant documentation and settings can be password protected.
In addition to the benefits features listed previously the wider range of XS balances also feature:
订购须知 OIML certified versions for all models in tables below, together with a full range of accessories for Excellence balances are also available - please see the website or contact Avantor.