Industrial scales, Excellence Plus XP series
供应商: METTLER TOLEDO
11130630 11130636 11130639 11130642 11130645 11130648 11130651 11130633
METB11130630EA 0 CNY
METB11130630 METB11130636 METB11130639 METB11130642 METB11130645 METB11130648 METB11130651 METB11130633
Industrial scales, Excellence Plus XP series
天平
The large platform versions of XP balances provide fast, stable results, safety and security, making the range particularly suitable where regulatory compliance is required for high capacity weighing. The design features MonoBloc HighSpeed technology, a large, colour, backlit touch screen that facilitates direct access to all functions and an extended range of applications that include density determinations, formulation, piece counting, statistics and dynamic weighing. Commercial keyboards or barcode readers can be easily connected with the PS/2 interface option or connectivity can be extended to networks with optional interfaces for Ethernet and Bluetooth®.
  • SmartScreen graphical colour display with touch screen operation for intuitive, easy operation with SmartSens, two sensors for hands-free balance operation
  • proFACT; fully automatic time and temperature controlled internal adjustment
  • QM toolbox includes user administration and password protection
  • MinWeigh reliably prevents minimal weight violations for full compliance and highest productivity
  • Level Control; warns the user if the balance is inclined and guides the user how to re-level the unit
  • Large stainless steel platform (IP54), with a removable control terminal
Certifications: OIML certified models are also available - please see the website or enquire for details.