Industrial scales, Excellence XS series
供应商: METTLER TOLEDO
11130660 11130663 11130666 11130669 11130672 11130675
METB11130660EA 0 CNY
METB11130660 METB11130663 METB11130666 METB11130669 METB11130672 METB11130675
Industrial scales, Excellence XS series
天平
The XS, large pan balances provide fast and accurate weighing using MonoBloc HighSpeed technology. The design also features a large, colour, backlit touch screen that facilitates direct access to all functions and an extended range of applications that include density determinations, formulation, piece counting, statistics and dynamic weighing. The units communicate in seven languages, and software can be downloaded from the internet.
  • Touch screen with alphanumeric keyboard for easy operation
  • Improved connectivity which includes Ethernet and Bluetooth options
  • DeltaTrac graphically displays amount of capacity utilised
  • FACT; fully automatic internal adjustment
  • Large stainless steel platform (IP 54), with a removable control terminal
Certifications: OIML certified models are available - please see the website or enquire for details. Also please contact Avantor for details of the full range of accessories for Excellence balances.