VWR®, Combined pH Electrodes with Internal Temperature Compensation
供应商: VWR Collection
VWRI662-0076EA 4125 CNY
VWRI662-0076 VWRI662-0077 VWRI662-0078 VWRI662-0080 VWRI662-0081 VWRI662-0083 VWRI662-0073 VWRI662-0075
VWR®, Combined pH Electrodes with Internal Temperature Compensation
电极 pH电极
Combined pH electrodes with 1 m fixed cable.
  • Three electrodes in one
  • Compatible with most pH meters
  • With protective cap for electrode preservation under optimal conditions
pH range: 0 to 14
Temperature range: 0 to +70 °C
Response time: 90% <1 second.