Tubing, rubber, vacuum
供应商: DEUTSCH NEUMANN
3020515 3020616 3020818 3021020 3020717 3020412 3021026 3020818A 3021222 3020820 3021228 3021531 3020309 3020513 3020715 3020824 3020614 3020618 3021016 3020412A 3020612 3021030 3020816A
DENE3020515EA 0 CNY
DENE3020515 DENE3020616 DENE3020818 DENE3021020 DENE3020717 DENE3020412 DENE3021026 DENE3020818A DENE3021222 DENE3020820 DENE3021228 DENE3021531 DENE3020309 DENE3020513 DENE3020715 DENE3020824 DENE3020614 DENE3020618 DENE3021016 DENE3020412A DENE3020612 DENE3021030 DENE3020816A
Tubing, rubber, vacuum
胶管 Transfer Tubing
Natural rubber, red.
  • Flexible, excellent tensile strength
  • Economical
Heavy wall, suitable for vacuum applications in the laboratory.
Temperature -resistance: −10 to +50 °C.
Hardness (Shore durometer A): 50.