DURAN® Test Tubes with or without Rim
供应商: DWK LIFE SCIENCES
261300302 261300602 261300808 261301307 261301607 261302106 261302303 261302603 261302809 261303308 261303608 261310106 261310303 261310603 261310809 261311102 261311308 261311608 261312604 261312801 261313309 261313609 261313806 261303805 261301101 261312107 261312304 261301204 261311205
SCOT261300302EA 0 CNY
SCOT261300302 SCOT261300602 SCOT261300808 SCOT261301307 SCOT261301607 SCOT261302106 SCOT261302303 SCOT261302603 SCOT261302809 SCOT261303308 SCOT261303608 SCOT261310106 SCOT261310303 SCOT261310603 SCOT261310809 SCOT261311102 SCOT261311308 SCOT261311608 SCOT261312604 SCOT261312801 SCOT261313309 SCOT261313609 SCOT261313806 SCOT261303805 SCOT261301101 SCOT261312107 SCOT261312304 SCOT261301204 SCOT261311205
DURAN® Test Tubes with or without Rim
管子 试验/取样管
DURAN®, borosilicate glass 3.3, clear. Thick-walled, exceptional mechanical strength.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes