试管
供应商: VWR Collection
VWRI212-1809EA 1316 CNY
VWRI212-1809 VWRI212-1823 VWRI212-1834 VWRI212-1837
试管
管子 试验/取样管
透明聚苯乙烯(PS).
  • Can be centrifuged up to 3000×g
  • Can be delivered sterile with red plug or non sterile without plug
  • Additional plugs and other colours available upon request
带通用试管,带圆底,应用范围广。