Strips of 8 PCR tubes with attached flat individual caps
供应商: BRAND
781332 781333
BRND781332EA 0 CNY
BRND781332 BRND781333
Strips of 8 PCR tubes with attached flat individual caps
管子 聚合酶链反应管
PP. Eight connected 0,2 or 0,15 ml PCR tubes with attached, flat individual caps.
  • Cap offers protection against contamination
  • Highly transparent cap makes them ideal for real-time PCR (qPCR)
  • Available in standard or low profile versions
  • Free from DNase, RNase and DNA
Colour: Clear