Individual PCR tubes with attached caps
供应商: BRAND
781300 781310 781305 781312 781301 781302 781303 781311 781313 781314 781304
BRND781300EA 0 CNY
BRND781300 BRND781310 BRND781305 BRND781312 BRND781301 BRND781302 BRND781303 BRND781311 BRND781313 BRND781314 BRND781304
Individual PCR tubes with attached caps
管子 聚合酶链反应管
PP. These 0,2 and 0,5 ml PCR tubes with attached caps are compatible with leading thermal cyclers with heated lids.
  • Unique cap design closes and opens easily without tools, yet ensures a tight fit to reduce sample evaporation
  • Flat caps allow simple labelling
  • Free from DNase, RNase and DNA
Colour: Clear