Strips of 8 PCR tubes and PCR caps
供应商: BRAND
781320 781340 781321 781322 781323 781324 781341 781342 781343 781344 781334 781325
BRND781320EA 0 CNY
BRND781320 BRND781340 BRND781321 BRND781322 BRND781323 BRND781324 BRND781341 BRND781342 BRND781343 BRND781344 BRND781334 BRND781325
Strips of 8 PCR tubes and PCR caps
管子 聚合酶链反应管
PP. Eight connected 0,2 ml tubes and domed or flat caps in strips of eight.
  • Flat caps are suitable for real-time PCR (qPCR)
  • Easy to open and close without tools
  • The tubes have a holding strap at one end, and the cover strips have a small lip on the side of each cap for careful, contamination-free opening
  • Free from DNase, RNase and DNA
Colour: Clear