高性能离心管
供应商: VWR Collection
89039-656 89039-660 89039-666
VWRI525-0402EA 0 CNY
VWRI525-0402 VWRU89039-656 VWRU89039-660 VWRU89039-666
高性能离心管
管子 离心管
PP材质,尖底,带紫色HDPE平盖或密封盖.
  • 经认证无细胞毒性,无核酸酶和无热原
  • 盖有两种形式:带平滑表面的平盖,或密封盖,可在使用摇床和水培养箱时,进行安全的密封的黑色刻度,易于读取,位于侧面的白色写入区域,可进行永久编码
  • 可用低至–80 °C的温度下存储和冷冻样品,可承受离心力达12500×g
These disposable, conical-bottom, free-standing tubes are extremely versatile, and can be used for routine tasks as well as high speed centrifuging. Rack-packed tubes are packed in freezable, recyclable, autoclavable racks. Tubes are sterilised by gamma irradiation (except free-standing tubes in bags or bulk). Tubes and caps are autoclavable and freezable.
Packaging: 包装:散装、袋装,或装在塑料的、可高温高压灭菌的支架上。