Laboratory trays
供应商: KARTELL
5701 5702 5703 5704 5705 5706 5707 5708 5709 5710 5711 5713 5714 5715
KART5701EA 0 CNY
KART5701 KART5702 KART5703 KART5704 KART5705 KART5706 KART5707 KART5708 KART5709 KART5710 KART5711 KART5713 KART5714 KART5715
Laboratory trays
盘子/皿/盆 实验室盘子/皿
PS, white.
  • Robust, impact resistant design
  • Heat resistant up to +70 °C
  • Smooth surfaces for easy cleaning
General purpose trays for a wide variety of laboratory uses. Also suitable for use in the food industry.