Laboratory storage trays, deep
供应商: KARTELL
542 543 544 602 603 604
KART542EA 0 CNY
KART542 KART543 KART544 KART602 KART603 KART604
Laboratory storage trays, deep
盘子/皿/盆 实验室盘子/皿
HDPE, white.
  • Robust, with lifting lugs and strengthening ribs
  • Stackable for space saving storage
  • 216-0263 has draining points on all sides - useful for defrosting samples
Multipurpose trays for a wide range of applications.