FluoroTrans® PVDF Transfer Membranes
供应商: CYTIVA PALL
BSP0161 BSP0157 BSP0159 PVM020C-099 PVM020C-195
PALLBSP0161EA 0 CNY
PALLBSP0161 PALLBSP0157 PALLBSP0159 PALLPVM020C-099 PALLPVM020C-195
FluoroTrans® PVDF Transfer Membranes
转移膜
Naturally hydrophobic PVDF, ideal for a wide variety of protein-analysis applications. The family of FluroTrans® media are white, microporous solid phase supports that bind proteins tenaciously via hydrophobic interactions. FluoroTrans® membrane is optimised for N-terminal protein sequencing. FluoroTrans® W membranes are optimised for Western transfer applications. They allow for sensitive protein detection with low background and very low protein burn-through. Immobilised proteins can be visually detected with Coomassie® Blue, Amido black, Ponceau S, and colloidal gold.
  • High protein binding capacity - typically absorbs 50% more protein than nylon or nitrocellulose
  • High tensile strength, so won't tear, crack or curl during handling, allowing easy removal of target bands for protein sequencing applications
  • High sensitivity for small peptides
Pore size: 0,2 µm
Typical thickness: 127 µm