通用温度计
供应商: VWR Collection
VWRI620-0857EA 2018.78 CNY
VWRI620-0857 VWRI620-0858 VWRI620-0860 VWRI620-1927 VWRI620-1928
通用温度计
温度计 Analogue Thermometers
该款温度计能够满足物理尺寸和精度的所有要求。
  • 在合适低温应用中可见的弯月面
  • 放大功能,便于读数
  • 白色背景,熔融刻度
Division: 1,0 °C
Accuracy: ±2,0 °C
Traceable to NIST/DKD. Compliant with ANSI/SAMA Z236.1-1983.
Certifications: NIST可追溯性。符合ANSI/SAMA Z236.1-1983标准。
Delivery information: Supplied with a multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability.
Packaging: 包装:Safety-Pak™独立包装。