Syringes, 3-piece
供应商: TERUMO
SS-10LE1 SS-02LE1 SS-05LE1 SS-50ES1 SS-50L1
TERUSS-10LE1EA 0 CNY
TERUSS-10LE1 TERUSS-02LE1 TERUSS-05LE1 TERUSS-50ES1 TERUSS-50L1
Syringes, 3-piece
注射器 医用注射器
Sterile, disposable, three-piece, latex-free.
  • Available with Luer slip tip or Luer lock threaded tip
  • Sterilised by electron beam irradiation
Certifications: Compliant with Directive 93/42/EEC Class IIa, organisation accredited by TÜV Rheinland (Germany), factory certified to ISO 9001 EN 46001, sterilisation process validated according to Standard NF EN 550.
Packaging: Individual sterile blister packs.