Microliter™ 800 series syringes
供应商: HAMILTON BONADUZ
84850 84851 84852 84853 84854 84855 84856 84857 84858 84859 84861 7642-01 CAL84851 CAL84854
HAMI84850EA 0 CNY
HAMI84850 HAMI84851 HAMI84852 HAMI84853 HAMI84854 HAMI84855 HAMI84856 HAMI84857 HAMI84858 HAMI84859 HAMI84861 HAMI7642-01 HAMICAL84851 HAMICAL84854
Microliter™ 800 series syringes
注射器 色谱用注射器
Universal injection syringes for GC. Similar to the 700 series but with a reinforced, protected plunger.
  • Extended handle simplifies sampling from test tubes or long necked flasks
  • Available with a standard 51 mm needle: Cemented (N) or removable (RN)