Gastight® 1700 series syringes
供应商: HAMILTON BONADUZ
201000 201050 201100 201150 201500 201600 202134 80000 80001 80008 80030 80039 80075 80200 80201 80208 80230 80239 80900 80901 80908 80920 80922 80930 80939 81000 81001 81008 81020 81022 81030 81039 81100 81101 81120 81122 81130 81139 81175 81201 81217 81220 81222 81230 81243 81227 7656-01 7655-01 7657-01 203566 7653-01 81127 7654-01 80011 80927 81026 81027 0159902 0160310 203560 203563 7658-01 80266 81041 81242 CAL80000 CAL80200 CAL80900 CAL81000 CAL81100 CAL81217 CAL81227 CAL7658-01 CAL81030 CAL81020 80085 80016 CAL81230 208639 CAL81220 208637 208641 208643
HAMI201000EA 0 CNY
HAMI201000 HAMI201050 HAMI201100 HAMI201150 HAMI201500 HAMI201600 HAMI202134 HAMI80000 HAMI80001 HAMI80008 HAMI80030 HAMI80039 HAMI80075 HAMI80200 HAMI80201 HAMI80208 HAMI80230 HAMI80239 HAMI80900 HAMI80901 HAMI80908 HAMI80920 HAMI80922 HAMI80930 HAMI80939 HAMI81000 HAMI81001 HAMI81008 HAMI81020 HAMI81022 HAMI81030 HAMI81039 HAMI81100 HAMI81101 HAMI81120 HAMI81122 HAMI81130 HAMI81139 HAMI81175 HAMI81201 HAMI81217 HAMI81220 HAMI81222 HAMI81230 HAMI81243 HAMI81227 HAMI7656-01 HAMI7655-01 HAMI7657-01 HAMI203566 HAMI7653-01 HAMI81127 HAMI7654-01 HAMI80011 HAMI80927 HAMI81026 HAMI81027 HAMI0159902 HAMI0160310 HAMI203560 HAMI203563 HAMI7658-01 HAMI80266 HAMI81041 HAMI81242 HAMICAL80000 HAMICAL80200 HAMICAL80900 HAMICAL81000 HAMICAL81100 HAMICAL81217 HAMICAL81227 HAMICAL7658-01 HAMICAL81030 HAMICAL81020 HAMI80085 HAMI80016 HAMICAL81230 HAMI208639 HAMICAL81220 HAMI208637 HAMI208641 HAMI208643
Gastight® 1700 series syringes
注射器 色谱用注射器
These syringes are the Gastight® version of the original 700 series syringes and are ideal for dispensing volumes from 1 µl up to 500 µl.
  • Fluid path includes stainless steel, borosilicate glass and PTFE
  • Gas sterilisable
Gastight® syringes are ideal for dispensing both liquids and gases and have a precision machined PTFE plunger tip which creates a leak free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Cemented needle (N): Cemented needles for low volume syringes are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Removable needle (RN): The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. This termination also allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Luer tip (LT): Luer tip syringes feature needles that are removable and fit over a ground glass hub which is tapered in the shape of a male Luer. This termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination also increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
Luer tip cemented needle (LTN): Luer tip cemented needle syringes are mid-volume syringes in which the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
PTFE Luer lock (TLL): PTFE Luer lock syringes have a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. This termination is also used with Hamilton diluters/dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 ml and greater syringes. Repeated autoclaving will shorten syringe life.
Needle point style 2: Sharp, beveled, curved, non-coring needle point recommended for septum penetration.
Needle point style 3: Blunt needle point for use with HPLC injection valves and for sample pipetting.
Needle point style 5: Conical needle with side port for penetration of septa, thin-gauged vinyls, and plastics without coring.
* For Microlab®
** Without slots