Stoppers, press down, VacuCap®
供应商: SIMPORT SCIENTIFIC
T402-13N T402-13R T402-13B T402-13G T402-13Y T402-13L T402-13GY T402-16N T402-16R T402-16B T402-16G T402-16Y T402-16L T402-16GY
SIMPT402-13NEA 0 CNY
SIMPT402-13N SIMPT402-13R SIMPT402-13B SIMPT402-13G SIMPT402-13Y SIMPT402-13L SIMPT402-13GY SIMPT402-16N SIMPT402-16R SIMPT402-16B SIMPT402-16G SIMPT402-16Y SIMPT402-16L SIMPT402-16GY
Stoppers, press down, VacuCap®
塞子 聚乙烯/低密度聚乙烯/高密度聚乙烯塞
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
  • Disposable seals are an inexpensive alternative to standard seals
  • Double flanged seal for an impermeable, secure fit
  • Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.