Magnetic stirring bars, tapered
供应商: COWIE
001.1915 001.1920 001.1925 001.1930 001.1935 001.1940 001.1950 001.1960 001.1970 001.1980 001.1910 001.1945
COWI001.1915EA 0 CNY
COWI001.1915 COWI001.1920 COWI001.1925 COWI001.1930 COWI001.1935 COWI001.1940 COWI001.1950 COWI001.1960 COWI001.1970 COWI001.1980 COWI001.1910 COWI001.1945
Magnetic stirring bars, tapered
搅拌器 磁力搅拌子
PTFE covered, strong Alnico V magnetic core. Smooth surface. Stirring action is similar to oval or pivot ring type stirring bars.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)