Cryo writing set, Nunc™
供应商: Thermo Fisher Scientific
339993 343850
NUNC339993EA 0 CNY
NUNC339993 NUNC343850
Cryo writing set, Nunc™
低温用马克笔
For labelling cryomaterials such as cryotubes, cryoboxes and cryolabels. Set contains one of each colour pen: black, blue, red and green.
  • Extremely durable ink
  • Fine tip
  • No colour change at low temperatures