Double spatulas, micro
供应商: BOCHEM
3340 3341 3342 3343 3344 3350 3351 3345
BOCH3340EA 0 CNY
BOCH3340 BOCH3341 BOCH3342 BOCH3343 BOCH3344 BOCH3350 BOCH3351 BOCH3345
Double spatulas, micro
样品勺
Stainless steel 18/10, anti-magnetic. Double spatulas for a wide range of uses.
  • One end with micro spoon or scoop and one end with micro spatula
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean