Spoon spatulas, micro
供应商: RSG
025.210 627.210 627.185 627.150 025.400 025.185 025.100 025.150 025.300
RSGA025.210EA 0 CNY
RSGA025.210 RSGA627.210 RSGA627.185 RSGA627.150 RSGA025.400 RSGA025.185 RSGA025.100 RSGA025.150 RSGA025.300
Spoon spatulas, micro
样品勺
Stainless steel 18/10, anti-magnetic.
  • With a micro spoon at one end and a micro spatula at the other
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean