Double spatulas, micro
供应商: BOCHEM
3300 3302 3303 3304 3306 3307 3301 3305
BOCH3300EA 0 CNY
BOCH3300 BOCH3302 BOCH3303 BOCH3304 BOCH3306 BOCH3307 BOCH3301 BOCH3305
Double spatulas, micro
样品勺
Stainless steel 18/10, anti-magnetic. With round handle, one flat and one scoop shaped, rounded spatula end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance