Pharmacist's spatulas
供应商: BOCHEM
3490 3491 3492 3493 3495 3496 3497 3494
BOCH3490EA 0 CNY
BOCH3490 BOCH3491 BOCH3492 BOCH3493 BOCH3495 BOCH3496 BOCH3497 BOCH3494
Pharmacist's spatulas
样品勺
Stainless steel blade, with wooden handle.
  • Blade with excellent heat resistance
  • Good chemical resistance
  • Quick and easy to clean