Sealing film, reflective, for PCR plates, Axygen®
供应商: Corning
PCR-AS-200 PCR-AS-600
AXYGPCR-AS-200EA 0 CNY
AXYGPCR-AS-200 AXYGPCR-AS-600
Sealing film, reflective, for PCR plates, Axygen®
密封件 微孔板封板膜和铝箔
This reflective aluminium film is ideal for sealing plates containing light sensitive samples. Seals during the thermo cycling process, when used in conjunction with a compression mat. Also provides an excellent seal when used with other polystyrene and polypropylene microplates.
  • Provides exceptional seal under the widest temperature conditions, −80 to +104 °C
  • Pierceable to allow for direct sample recovery without peeling
  • Proprietary adhesive is inert and chemically resistant to DMSO