Breatheable sealing film seals, Axygen®
供应商: Corning
BF-400 BF-400-S
AXYGBF-400EA 0 CNY
AXYGBF-400 AXYGBF-400-S
Breatheable sealing film seals, Axygen®
密封件 微孔板封板膜和铝箔
Porous film seals PP and PS culture plates, 96 and 384-well plates, and other assay plates.
  • Allows effective gas exchange for cellular and bacterial cultivation and prevents contamination
  • Sterile packs available