Quick-release clamps for flat flange connections, DURAN®
供应商: DWK LIFE SCIENCES
290713407 290714609 290715108 290715708 290716104
SCOT290713407EA 0 CNY
SCOT290713407 SCOT290714609 SCOT290715108 SCOT290715708 SCOT290716104
Quick-release clamps for flat flange connections, DURAN®
夹具 通用夹具
Stainless steel.
  • Simple assembly and disassembly
  • With three holding segments
  • Quick release
For clamping reaction vessels and lids together.