O-rings for flat flanges, DURAN®
供应商: DWK LIFE SCIENCES
292254602 292255701 292256106 292255101 292253409
SCOT292254602EA 0 CNY
SCOT292254602 SCOT292255701 SCOT292256106 SCOT292255101 SCOT292253409
O-rings for flat flanges, DURAN®
密封件 Sealing Rings
Silicone (VMQ), transparent.
  • Guarantee excellent joint seal
  • Heat resistant from −50 to +200 °C
  • Good chemical resistance
O-rings for flat flanges with groove.