Flat bottom chambers
供应商: CAMAG
022.5150 022.5151 022.5250 022.5257 022.5259
CAMA022.5150EA 0 CNY
CAMA022.5150 CAMA022.5151 CAMA022.5250 CAMA022.5257 CAMA022.5259
Flat bottom chambers
薄层色谱展开槽
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.