Serological pipettes, total delivery
供应商: BRAND
27109 27107 27113 27115 27112
BRND27109EA 0 CNY
BRND27109 BRND27107 BRND27113 BRND27115 BRND27112
Serological pipettes, total delivery
移液管和移液器 血清移液管
AR-Glas®.
  • Calibrated to deliver 'Ex'
  • Graduations and inscriptions in ETERNA amber stain, short graduation marks
  • Cotton plugged upper end
Tip Ø: 2 mm for 1 and 2 ml pipettes; 3 mm for 5, 10 and 25 ml pipettes.