Micro pipettes, ringcaps®
供应商: HIRSCHMANN
9600122 9600105 9600110 9600120 9600125 9600140 9600144 9600150 9600199
HIRS9600122EA 0 CNY
HIRS9600122 HIRS9600105 HIRS9600110 HIRS9600120 HIRS9600125 HIRS9600140 HIRS9600144 HIRS9600150 HIRS9600199
Micro pipettes, ringcaps®
移液管和移液器 微血管
DURAN®, borosilicate glass, with ring mark, disposable.
  • Colour coded according to ISO for clear identification
  • Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
Certifications: ISO 9000, DIN ISO 7550, IVD 98/79 EC