Graduated pipettes
供应商: BRAND
27607 27610 27613 27614
BRND27607EA 0 CNY
BRND27607 BRND27610 BRND27613 BRND27614
Graduated pipettes
移液管和移液器 刻度移液管
PP, high clarity.
  • Calibrated to deliver (TD, Ex)
  • Circular markings and inscriptions in blue
  • Cleaning temperature below 60 °C recommended in order to preserve markings and inscriptions
Suction tube outer Ø: 8 mm (except 612-4915: 10 mm)