FTA® reagent and accessories, Whatman™
供应商: QIAGEN
WB120204 WB100003 WB100032 WB100035 WB100036 WB100037
QIAGWB120204EA 0 CNY
QIAGWB120204 QIAGWB100003 QIAGWB100032 QIAGWB100035 QIAGWB100036 QIAGWB100037
FTA® reagent and accessories, Whatman™
核酸储存系统
For collection, storage, processing and shipping FTA® cards.
  • FTA® purification reagent: For purification of nucleic acids stored on FTA® cards
  • Ensures superior quality DNA for PCR or SNP analysis
  • Removes haem, PCR inhibitors and other potential contaminants
  • Non toxic, hypoallergenic, aqueous solution
Sterile foam tipped applicator: For the collection of saliva and buccal cells. Non abrasive foam head is the same size as sample area on indicating FTA® Cards to facilitate sample application.
Sterile omni swab: Non invasive device for collecting saliva and cheek buccal cells. Brush-like swab head easily ejects from the stem of the swab for transfer of samples into tubes or multiwell plates. Pre-sterilised and individually wrapped for single use.
Multi-barrier pouches: For storing and transporting FTA® cards
Seven laminated layers protect the card from exposure to gas or liquid contamination
Tamper-evident seal maintains sample security
Outer paper surface for labelling or writing
Recommended for the precise punching of FTA® Cards. No sample carryover when recommended procedures are used. Tips provide up to 2000 punches. Polished steel tip is case hardened and can be sterilised. The cutting mat ensures clean sample cuts and extends the life of the cutting tip.