CloneSaver™ Cards, Whatman™
供应商: QIAGEN
WB120028 WB100024
QIAGWB120028EA 0 CNY
QIAGWB120028 QIAGWB100024
CloneSaver™ Cards, Whatman™
核酸储存系统
FTA® technology in 96-well format for high throughput applications.
  • Bacteriophages are inactivated
  • DNA is easily accessible for downstream applications, such as transformation, PCR® or sequencing
  • Up to 96 samples can be stored on each card
Designed for the collection, long-term storage and purification of plasmid and BAC DNA in a 96-well format. Bacterial cultures, resuspended colonies or glycerol stock are applied, cells are lysed, and plasmid or BAC DNA is stabilised for long-term storage or immediate processing.