Deep well plates and cover mats
供应商: BRAND
701350 701354 701352 701346 701355 701340 701342
BRND701350EA 0 CNY
BRND701350 BRND701354 BRND701352 BRND701346 BRND701355 BRND701340 BRND701342
Deep well plates and cover mats
微孔板 深孔微孔板
PP or PS, non sterile. Designed for a wide range of applications, such as high throughput screening, cell and tissue cultures, serial dilutions, reagent transfer, and sample storage down to −80 °C (PP) or −20 °C (PS). Deep well plates are manufactured in standard ANSI/SLAS 1 and 4 format and are stackable for easy storage.
  • PP for high chemical resistance, for example, against DMSO, phenol, chloroform
  • Autoclavable at 121 °C (2 bar) acc. DIN EN 285 (PP plates only)
  • Round shape wells allow optimal sample mixing and recovery
  • Alphanumeric coding and cut-away corner simplify sample identification and orientation
  • Can be used with multi channel pipettes and automatic liquid handling systems from all leading manufacturers
  • Free from DNA, DNase and RNase (except low profile plate which is not tested)