Optical bottom plates, 96- and 384-well, Nunc™
供应商: Thermo Fisher Scientific
142761 142762 242763 242764 265301 265302 240074 164586 265300
NUNC142761EA 0 CNY
NUNC142761 NUNC142762 NUNC242763 NUNC242764 NUNC265301 NUNC265302 NUNC240074 NUNC164586 NUNC265300
Optical bottom plates, 96- and 384-well, Nunc™
微孔板 高温超导微孔板
PS upper structure with either PS film or no. 1.5 coverglass base. Designed for fluorescence and luminescence based assays, these plates have white or black upper structures with optically clear bottoms. Consequently, they can be read from bottom and top, or both.
  • Compatible with automated handling equipment
  • Alphanumeric well identification
  • Optimised for scintillation counting
  • Low fluorescence background ensures accurate results, due to higher signal-to-noise ratios