Measuring cylinders, low form, class B, borosilicate glass
供应商: HIRSCHMANN
2400160 2400170 2400175 2400180 2400186
HIRS2400160EA 0 CNY
HIRS2400160 HIRS2400170 HIRS2400175 HIRS2400180 HIRS2400186
Measuring cylinders, low form, class B, borosilicate glass
量筒 Measuring Cylinders
DURAN®, borosilicate glass, low form, class B.
  • Calibrated to contain 'In'
  • Hexagonal base with spout
  • Marks and inscriptions in amber stain, short graduation marks
DIN EN ISO 4788