Stools, mobile, STEP
供应商: WEDO WERNER DORSCH
212200 212212 212237 212202 212203
WEDO212200EA 0 CNY
WEDO212200 WEDO212212 WEDO212237 WEDO212202 WEDO212203
Stools, mobile, STEP
梯子和踏凳
These robust kick stools are manufactured from a recyclable UV resistant PP and mounted on three castors allowing complete mobility. When in use the castors retract, providing a secure platform.
  • Supports up to 150 kg
  • Snap-Lock assembly, no tools required
Certifications: EN 14183 (DIN 4569), TÜV GS approved.