Biopsy processing/embedding cassettes, Histosette® I, M491
供应商: SIMPORT SCIENTIFIC
M491-2 M491-3 M491-4 M491-5 M491-6 M491-9 M491-10 M491-11 M491-12 M491-7 M491-8
SIMPM491-2EA 0 CNY
SIMPM491-2 SIMPM491-3 SIMPM491-4 SIMPM491-5 SIMPM491-6 SIMPM491-9 SIMPM491-10 SIMPM491-11 SIMPM491-12 SIMPM491-7 SIMPM491-8
Biopsy processing/embedding cassettes, Histosette® I, M491
包埋盒 活检处理盒
Acetal polymer.
  • Resistant to all commonly used histological solvents
  • Square openings (1 mm) to maximise fluid exchange and ensure proper drainage
  • Integral lid
  • Anterior writing area at a 30° angle