Biopsy processing/embedding cassettes, Histosette® II, M493
供应商: SIMPORT SCIENTIFIC
M493-2 M493-4 M493-5 M493-6 M493-7 M493-8 M493-9 M493-10 M493-11 M493-12 M493-3
SIMPM493-2EA 0 CNY
SIMPM493-2 SIMPM493-4 SIMPM493-5 SIMPM493-6 SIMPM493-7 SIMPM493-8 SIMPM493-9 SIMPM493-10 SIMPM493-11 SIMPM493-12 SIMPM493-3
Biopsy processing/embedding cassettes, Histosette® II, M493
包埋盒 活检处理盒
Acetal polymer.
  • No need for biopsy pads
  • Resistant to most commonly used histological solutions
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems