Biopsy processing/embedding cassettes, Microsette™ M503
供应商: SIMPORT SCIENTIFIC
M503-8 M503-12 M503-10 M503-11 M503-2 M503-3 M503-4 M503-5 M503-6 M503-7 M503-9
SIMPM503-8EA 0 CNY
SIMPM503-8 SIMPM503-12 SIMPM503-10 SIMPM503-11 SIMPM503-2 SIMPM503-3 SIMPM503-4 SIMPM503-5 SIMPM503-6 SIMPM503-7 SIMPM503-9
Biopsy processing/embedding cassettes, Microsette™ M503
包埋盒 活检处理盒
Acetal polymer.
  • No need for biopsy pads
  • Six compartments, each 7×12 mm, numbered 1 to 6
  • 2000 square openings (each 0,26 mm) to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems